Invitrogen
Manufacturer Code:MA119605
Catalog # 01-676-333
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HI149 |
Immunogen | Human thymocytes. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD1 FCB6 HTA1 R4 T6 CD1A antigen a polypeptide T-cell surface antigen T6/Leu-6 T-cell surface glycoprotein CD1a cluster of differentiation 1 A cortical thymocyte antigen CD1A; CD1a; CD1A antigen, a polypeptide; cluster of differentiation 1 A; cortical thymocyte antigen CD1A; differentiation antigen CD1-alpha-3; epidermal dendritic cell marker CD1a; hTa1 thymocyte antigen; T-cell surface antigen T6/Leu-6; T-cell surface glycoprotein CD1a
Gene Aliases: CD1; CD1A; FCB6; HTA1; R4; T6
UniProt ID: (Human) P06126
Entrez Gene ID: (Human) 909
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.