Invitrogen
Manufacturer Code:17197163
Catalog # 50-151-15
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 4B12 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Bukitt&primes lymphoma receptor 2 C-C chemokine receptor type 7 CC chemokine receptor 7 CCR-7 CD197 Chemokine (C-C Motif) Receptor 7 chemokine (C-C) receptor 7 CMKBR7 EBI1 EBV-induced G protein-coupled receptor 1 EBV-induced G-protein coupled receptor 1 Epstein-Barr virus induced gene 1 Epstein-Barr Virus-Induced G-Protein Coupled Receptor 1 lymphocyte-specific G protein-coupled peptide receptor MIP-3 Beta Receptor; C-C chemokine receptor type 7; C-C CKR-7; CD197; chemokine (C-C) receptor 7; EBI1; EBV-induced G-protein coupled receptor 1; Epstein-Barr virus-induced G-protein coupled receptor 1; MIP-3 beta receptor
Gene Aliases: CC-CKR-7; CCR-7; Ccr7; CD197; Cdw197; Cmkbr7; EBI1; Ebi1h
UniProt ID: (Mouse) P47774
Entrez Gene ID: (Mouse) 12775
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.