Invitrogen
Manufacturer Code:50719682
Catalog # 50-112-8902
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | sirx6 |
Conjugate | eFluor 660 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: BN-1 CKR-L3 CKR6 CKRL3 CMKBR6 DCR2 DRY-6 GPR-CY4 GPR29 GPRCY4 STRL22; C-C chemokine receptor type 6; C-C CKR-6; CD196; KY411
Gene Aliases: CC-CKR-6; CCR-6; Ccr6; Cmkbr6; KY411
UniProt ID: (Mouse) O54689
Entrez Gene ID: (Mouse) 12458
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.