Invitrogen
Manufacturer Code:165911025
Catalog # 16-591-1025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | MFL3 |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: ADAM10-processed FasL form; ADAM10-processed FasL form ALPS1B APL apoptosis (APO-1) antigen ligand 1 Apoptosis (APO-1) antigen ligand 1 (Fas antigen ligand) apoptosis antigen ligand APT1LG1 APTL CD178 CD95 ligand CD95L CD95-L CD95L protein fas antigen ligand Fas ligand Fas ligand (TNF superfamily member 6) Fasl Fas-L FasL ICD FasL intracellular domain Faslg Fas-LG generalized lymphoproliferative disease gld mutant tumor necrosis factor family member 6 Receptor-binding FasL ectodomain sFasL Soluble Fas ligand SPA SPPL2A-processed FasL form TNFL Tnfsf6 TNLG1A Tumor necrosis factor (ligand) superfamily member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand) Tumor necrosis factor (ligand) superfamily member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand) Tumor necrosis factor ligand tumor necrosis factor ligand 1A tumor necrosis factor ligand superfamily member 6 Tumor necrosis factor ligand superfamily member 6 membrane form Tumor necrosis factor ligand superfamily member 6 soluble form; APL; CD178; CD95 ligand; CD95-L; Fas antigen ligand; Fas ligand; FasL ICD; FasL intracellular domain; generalized lymphoproliferative disease; Receptor-binding FasL ectodomain; sFasL; Soluble Fas ligand; SPA; SPPL2A-processed FasL form; Tumor necrosis factor ligand superfamily member 6; Tumor necrosis factor ligand superfamily member 6, membrane form; Tumor necrosis factor ligand superfamily member 6, soluble form
Gene Aliases: APT1LG1; CD178; CD95-L; CD95L; Fas-L; Fasl; Faslg; gld; Tnfsf6
UniProt ID: (Mouse) P41047
Entrez Gene ID: (Mouse) 14103
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.