Invitrogen
Manufacturer Code:PA532400
Catalog # PIPA532400
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from near N-terminus of human FAS ligand. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAM10-processed FasL form; APL; apoptosis (APO-1) antigen ligand 1; Apoptosis (APO-1) antigen ligand 1 (Fas antigen ligand); Apoptosis antigen ligand; APTL; CD178; CD95 ligand; CD95-L; CD95L protein; Fas antigen ligand; Fas ligand; Fas ligand (TNF superfamily, member 6); FasL apoptosis antigen ligand 1 CD95 ligand CD95L CD178 APO1 Fas Ligand Fas-LG; FasL ICD; FasL intracellular domain; generalized lymphoproliferative disease; mutant tumor necrosis factor family member 6; Receptor-binding FasL ectodomain; sFasL; Soluble Fas ligand; SPA; SPPL2A-processed FasL form; Tumor necrosis factor (ligand) superfamily member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand); Tumor necrosis factor (ligand) superfamily, member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand); tumor necrosis factor ligand 1A; Tumor necrosis factor ligand superfamily member 6; Tumor necrosis factor ligand superfamily member 6, membrane form; Tumor necrosis factor ligand superfamily member 6, soluble form
Gene Aliases: ALPS1B; APT1LG1; APTL; CD178; CD95-L; CD95L; Fas-L; FASL; FASLG; gld; TNFSF6; TNLG1A
UniProt ID: (Human) P48023, (Mouse) P41047, (Rat) P36940
Entrez Gene ID: (Human) 356, (Mouse) 14103, (Rat) 25385
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.