Invitrogen
Manufacturer Code:MA529554
Catalog # PIMA529554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 31 |
Immunogen | Recombit Human PSGL-1/CD162/SELPLG Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD162; CD162 CLA PSGL-1 PSGL1 P-selectin glycoprotein ligand 1 cutaneous lymphocyte-associated associated antigen SELPG P-Selectin Glycoprotein Ligand 1; cutaneous lymphocyte-associated associated antigen; P-selectin glycoprotein ligand 1; PSGL-1; Selectin P ligand
Gene Aliases: CD162; CLA; PSGL-1; PSGL1; SELPLG
UniProt ID: (Human) Q14242
Entrez Gene ID: (Human) 6404
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.