Invitrogen
Manufacturer Code:PA5102457
Catalog # PIPA5102457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SELPLG corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD162; CD162 CLA cutaneous lymphocyte-associated associated antigen leukocyte cell surface adhesion molecule P-selectin glycoprotein ligand 1 P-selectin glycoprotein ligand 1 propeptide P-selectin glycoprotein ligand-1 Psgl1 PSGL-1 Selectin P ligand selectin platelet (p-selectin) ligand Selp1 SELPG Selpl SELPLG; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; PSGL-1; Selectin P ligand; selectin, platelet (p-selectin) ligand
Gene Aliases: CD162; CLA; PSGL-1; PSGL1; Selpl; SELPLG
UniProt ID: (Human) Q14242
Entrez Gene ID: (Human) 6404, (Rat) 363930
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.