Invitrogen
Manufacturer Code:14160185
Catalog # 50-129-32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBioCNX46-3 (CNX46-3) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD160; CD160 antigen; CD160 antigen, soluble form; natural killer cell BY55; Natural killer cell receptor BY55; NK1 NK28 CD160 antigen CD160 transmembrane isoform CD160-delta Ig natural killer cell receptor BY55 natural killer cell receptor
Gene Aliases: AU045688; By55; Cd160
UniProt ID: (Mouse) O88875
Entrez Gene ID: (Mouse) 54215
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.