Invitrogen
Manufacturer Code:MA517968
Catalog # PIMA517968
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CNX46-3 |
Immunogen | Mouse CD160 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD160; CD160 antigen; CD160 antigen, soluble form; natural killer cell BY55; Natural killer cell receptor BY55; NK1 NK28 CD160 antigen CD160 transmembrane isoform CD160-delta Ig natural killer cell receptor BY55 natural killer cell receptor
Gene Aliases: AU045688; By55; Cd160
UniProt ID: (Mouse) O88875
Entrez Gene ID: (Mouse) 54215
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.