R&D Systems
Manufacturer Code:MAB38991
Catalog # MAB38991
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 342705 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse CD160 Gly28-Ser160 Accession # AAH21596 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: BY55 BY55FLJ46513 CD160 CD160 antigenimmunoglobulin superfamily member CD160 molecule Natural killer cell receptor BY55 NK1 NK28; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 transmembrane isoform; CD160-delta Ig; Natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member
Gene Aliases: BY55; CD160; NK1; NK28
UniProt ID: (Human) O95971
Entrez Gene ID: (Human) 11126
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.