Novus Biologicals
Manufacturer Code:NBP290074100UL
Catalog # NB021611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 275 |
Immunogen | Recombinant Human CD160 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: BY55FLJ46513 CD160 antigenimmunoglobulin superfamily member CD160 molecule Natural killer cell receptor BY55; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 transmembrane isoform; CD160-delta Ig; Natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member
Gene Aliases: BY55; CD160; NK1; NK28
UniProt ID: (Human) O95971
Entrez Gene ID: (Human) 11126
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.