Invitrogen
Manufacturer Code:48016180
Catalog # 50-162-62
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 93 |
Conjugate | eFluor 450 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD32; Fc gamma receptor IIB; Fc gamma RIIB; Fc-gamma RII; Fc-gamma-RII; fc-gamma-RIIB; FcgammaRIIB; FCGR3 FCGR3A FCGR3B CD16A CD16B Fc gamma Receptor III Fc gamma Receptor 3 RP11-5K23.1 FCGR3 FCGRIII FCR-10 FCRIII FCRIIIA FCGR2 CD32 Fc gamma Receptor II CD32 receptor 2 Fc gamma Receptor 2 FCGR2A FCGR2B FCGR2C Low affinity immunoglobulin gamma Fc region receptor II-b CDw32 Fc-gamma RII-b Fc-gamma-RIIb FcRII-b FCG2 Low affinity immunoglobulin gamma Fc region receptor II-c Fc-gamma RII-c Fc-gamma-RIIc FcRII-c FGFR2B; IgG Fc receptor II beta; Low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; Ly-17; Lymphocyte antigen 17
Gene Aliases: AI528646; CD32; F630109E10Rik; Fc[g]RII; Fcgr2; Fcgr2a; Fcgr2b; FcgRII; Fcr-2; Fcr-3; fcRII; Ly-17; Ly-m20; LyM-1
UniProt ID: (Mouse) P08101
Entrez Gene ID: (Mouse) 14130
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.