Invitrogen
Manufacturer Code:12158941
Catalog # 50-106-71
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HP-MA4 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD158 antigen-like family member A; CD158 antigen-like family member G; CD158 antigen-like family member H; CD158a; CD158g; CD158h; Killer cell immunoglobulin-like receptor 2DL1; Killer cell immunoglobulin-like receptor 2DS1; Killer cell immunoglobulin-like receptor 2DS5; killer cell immunoglobulin-like receptor KIRDS1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; killer Ig receptor; killer inhibitory receptor 2-2-1; killer-cell immunoglobulin-like receptor; MHC class I NK cell receptor; MHC class I NK cell receptor Eb6 ActI; natural killer cell inhibitory receptor; Natural killer-associated transcript 1; Natural killer-associated transcript 9; NKAT-1; NKAT-9; p58 killer cell inhibitory receptor KIR-K64; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-42/47.11; p58.1 MHC class-I-specific NK receptor
Gene Aliases: CD158A; CD158G; CD158H; KIR-K64; KIR221; KIR2DL1; KIR2DP1DL1; KIR2DS1; KIR2DS4; KIR2DS5; NKAT; NKAT-1; NKAT1; NKAT9; p50.1; p58.1
UniProt ID: (Human) P43626, (Human) Q14954, (Human) Q14953
Entrez Gene ID: (Human) 3802, (Human) 3806, (Human) 3810
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.