Novus Biologicals
Manufacturer Code:NB6001241
Catalog # NB6001241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | D171 |
Immunogen | HeLa cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD155; CD155 CD155 antigen HVED NECL5 NECL-5 nectin-like 5 Nectin-like protein 5 poliovirus receptor PVSFLJ25946 Tage4; NECL-5; Nectin-like protein 5; Poliovirus receptor
Gene Aliases: CD155; HVED; Necl-5; NECL5; PVR; PVS; TAGE4
UniProt ID: (Human) P15151
Entrez Gene ID: (Human) 5817
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.