R&D Systems
Manufacturer Code:FAB6909G
Catalog # FAB6909G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 690912 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse CD155/PVR Asp29-Leu348 Accession # NP_081790 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD155; CD155 CD155 antigen HVED Necl-5 NECL5 nectin-like 5 Nectin-like protein 5 poliovirus receptor PVR PVS PVSFLJ25946 Tage4; NECL-5; Nectin-like protein 5; Poliovirus receptor
Gene Aliases: CD155; HVED; Necl-5; NECL5; PVR; PVS; TAGE4
UniProt ID: (Human) P15151
Entrez Gene ID: (Human) 5817
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.