Invitrogen
Manufacturer Code:MA513490
Catalog # PIMA513490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | D171 |
Immunogen | HeLa cells |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD155; NECL-5; nectin-like 5 nectin-like protein 5 PVR PVS; Nectin-like protein 5; Poliovirus receptor; tumor-associated antigen 1
Gene Aliases: 3830421F03Rik; CD155; D7Ertd458e; HVED; mE4; Necl-5; NECL5; PVR; PVS; Taa1; TAGE4
UniProt ID: (Human) P15151
Entrez Gene ID: (Human) 5817, (Mouse) 52118
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.