Invitrogen
Manufacturer Code:MA529762
Catalog # PIMA529762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 305 |
Immunogen | Recombit Mouse CD155/PVR protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD112; Herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; mHveB; Murine herpes virus entry protein B; murine herpesvirus entry protein B; Nectin cell adhesion molecule 2; Nectin-2; nectin-like 5 nectin-like protein 5 PVR PVS; Poliovirus receptor homolog; Poliovirus receptor-related protein 2; poliovirus sensitivity
Gene Aliases: AI325026; AI987993; Cd112; MPH; nectin-2; Nectin2; Pvr; Pvrl2; Pvs
UniProt ID: (Mouse) P32507
Entrez Gene ID: (Mouse) 19294
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.