Invitrogen
Manufacturer Code:MA133895
Catalog # PIMA133895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | 39H5 |
Immunogen | Proprietary immunogen - Contact ABR for details. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: CD154; CD40 ligand; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40-L; RP23-153G22.3 CD154 CD40-L Cd40l HIGM1 IGM IMD3 Ly-62 Ly62 T-BAM TRAP Tnfsf5 gp39 T-cell antigen Gp39 TNF-related activation protein tumor necrosis factor ligand superfamily member 5; sCD40L; T-cell antigen Gp39; TNF-related activation protein; TRAP; Tumor necrosis factor ligand superfamily member 5
Gene Aliases: CD154; CD40-L; Cd40l; Cd40lg; gp39; HIGM1; IGM; IMD3; Ly-62; Ly62; T-BAM; Tnfsf5; TRAP
UniProt ID: (Mouse) P27548
Entrez Gene ID: (Mouse) 21947
Molecular Function:
cytokine
signaling molecule
tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.