Invitrogen
Manufacturer Code:PA578983
Catalog # PIPA578983
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse CD40L recombit protein (Position: E107-L260). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand CD40-L T-cell antigen Gp39 TNF-related activation protein TRAP Tumor necrosis factor ligand superfamily member 5 CD154 CD40 ligand membrane form CD40 ligand soluble form Cd40lg Cd40l Tnfsf5; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40-L; sCD40L; T-B cell-activating molecule; T-cell antigen Gp39; TNF-related activation protein; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; Tumor necrosis factor ligand superfamily member 5
Gene Aliases: CD154; CD40-L; CD40L; CD40LG; gp39; hCD40L; HIGM1; IGM; IMD3; Ly-62; Ly62; T-BAM; TNFSF5; TRAP
UniProt ID: (Human) P29965, (Mouse) P27548, (Rat) Q9Z2V2
Entrez Gene ID: (Human) 959, (Mouse) 21947, (Rat) 84349
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.