Invitrogen
Manufacturer Code:12150180
Catalog # 50-106-42
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 9D1 |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD150; CD150 CDw150 SLAM IPO-3 signaling lymphocytic activation molecule SLAMF1; signaling lymphocyte activation molecule; Signaling lymphocytic activation molecule; SLAM family member 1
Gene Aliases: 4933415F16; AA177906; CD150; CDw150; ESTM51; IPO-3; Slam; Slamf1
UniProt ID: (Mouse) Q9QUM4
Entrez Gene ID: (Mouse) 27218
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
kinase
protein kinase
receptor
signaling molecule
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.