Invitrogen
Manufacturer Code:MA531201
Catalog # PIMA531201
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 110 |
Immunogen | Recombit Human CD150/SLAM/SLAMF1 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD150; CD150 CDw150 SLAM IPO-3 signaling lymphocytic activation molecule SLAMF1; CDw150; IPO-3; Signaling lymphocytic activation molecule; SLAM family member 1
Gene Aliases: CD150; CDw150; SLAM; SLAMF1
UniProt ID: (Human) Q13291
Entrez Gene ID: (Human) 6504
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
kinase
protein kinase
receptor
signaling molecule
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.