Novus Biologicals
Manufacturer Code:NB500376PE
Catalog # NB500376PE
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MEM-158 |
Immunogen | Human granulocytes |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
For Research Use Only
Protein Aliases: alpha (1,3) fucosyltransferase, myeloid-specific; Alpha-(1,3)-fucosyltransferase 4; CD 15 CD-15 Lewis X LeX SSEA-1 stage-specific embryonic antigen 1; ELAM ligand fucosyltransferase; ELAM-1 ligand fucosyltransferase; Fuc-TIV; Fucosyltransferase 4; fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific); Fucosyltransferase IV; fucT-IV; Galactoside 3-L-fucosyltransferase; Lewis X; stage-specific embryonic antigen 1
Gene Aliases: CD15; ELFT; FCT3A; FUC-TIV; FUT4; FUTIV; LeX; SSEA-1
UniProt ID: (Human) P22083
Entrez Gene ID: (Human) 2526
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.