Invitrogen
Manufacturer Code:MA511789
Catalog # PIMA511789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MMA (LeuM1) |
Immunogen | U937 histiocytic cell line |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD15 ELFT FCT3A Alpha-(13)-fucosyltransferase 4 ELAM-1 ligand fucosyltransferase Fucosyltransferase 4 Fucosyltransferase IV Fuc-TIV FucT-IV Galactoside 3-L-fucosyltransferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.