Invitrogen
Manufacturer Code:MA112651
Catalog # 01-674-032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MCS-1 |
Immunogen | White blood cells. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD15 ELFT FCT3A FUC-TIV FUTIV LeX SSEA-1
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.