Proteintech
Manufacturer Code:119891AP150UL
Catalog # 50-172-7620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CD147 Fusion Protein Ag2619 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5A11/Basigin; 5F7; 5F7 Basigin basigin (Ok blood group) BSG CD147 Collagenase stimulatory factor EMMPRIN HAb18G M6 OK OK blood group antigen TCSF; Basic immunoglobulin superfamily; Basigin; CD147; Collagenase stimulatory factor; EMMPRIN; Extracellular matrix metalloproteinase inducer; gp 42; HAb18G; Hepatoma-associated antigen; HT7 antigen; Leukocyte activation antigen M6; Membrane glycoprotein gp42; neurothelin; OK blood group antigen; TCSF; Tumor cell-derived collagenase stimulatory factor
Gene Aliases: 5F7; AI115436; AI325119; BSG; CD147; EMMPRIN; HT-7; OK; TCSF; UNQ6505/PRO21383
UniProt ID: (Human) P35613, (Mouse) P18572
Entrez Gene ID: (Human) 682, (Mouse) 12215
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.