Invitrogen
Manufacturer Code:17142941
Catalog # 50-245-666
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HTF-1 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD142; Coagulation factor III; coagulation factor III (thromboplastin, tissue factor); F3 Coagulation factor 3 TF TFA; TF; Thromboplastin; Tissue factor
Gene Aliases: CD142; F3; TF; TFA
UniProt ID: (Human) P13726
Entrez Gene ID: (Human) 2152
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.