Invitrogen
Manufacturer Code:MA524102
Catalog # PIMA524102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 355220 |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Coagulation Factor III Ala29-Glu251 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD142; Coagulation factor III; F3 Coagulation factor 3 TF TFA Coagulation Factor III; TF; Tissue factor
Gene Aliases: AA409063; CD142; Cf-3; Cf3; F3; TF
UniProt ID: (Mouse) P20352
Entrez Gene ID: (Mouse) 14066
Molecular Function:
cytokine receptor
defense/immunity protein
receptor
type I cytokine receptor
type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.