Invitrogen
Manufacturer Code:PA579227
Catalog # PIPA579227
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived mouse Tissue Factor recombit protein (Position: A29-A294). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD142; Coagulation factor III; TF; Tissue factor; Tissue factor TF Coagulation factor III CD142 F3 Cf-3 Cf3
Gene Aliases: AA409063; CD142; Cf-3; Cf3; F3; TF
UniProt ID: (Mouse) P20352
Entrez Gene ID: (Mouse) 14066
Molecular Function: cytokine receptor defense/immunity protein receptor type I cytokine receptor type II cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.