Invitrogen
Manufacturer Code:MA512400
Catalog # PIMA512400
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MI15 |
Immunogen | U-266 cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; SDC1 SYND1 SDC CD antigen 138; SYND1; syndecan proteoglycan 1; Syndecan-1
Gene Aliases: CD138; HSPG; SDC; SDC1; SYND1; SYNDECA; Syndecan
UniProt ID: (Human) P18827, (Rat) P26260
Entrez Gene ID: (Human) 6382, (Rat) 25216
Molecular Function:
cell adhesion molecule
cytoskeletal protein
defense/immunity protein
extracellular matrix glycoprotein
extracellular matrix protein
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.