Invitrogen
Manufacturer Code:PA516918
Catalog # PIPA516918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encoding aa 18-218 human CD138 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; SDC1 SYND1 SDC CD antigen 138; SYND1; syndecan proteoglycan 1; Syndecan-1; synstatin
Gene Aliases: AA408134; AA409076; CD138; SDC; SDC1; Sstn; syn-1; Synd; Synd-1; SYND1; syndecan
UniProt ID: (Human) P18827, (Mouse) P18828
Entrez Gene ID: (Human) 6382, (Mouse) 20969
Molecular Function: cell adhesion molecule cytoskeletal protein defense/immunity protein extracellular matrix glycoprotein extracellular matrix protein membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.