Invitrogen
Manufacturer Code:MA535974
Catalog # PIMA535974
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | A24 |
Immunogen | Recombinant Human CD137 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 4 1 BB 41BB 4-1BB 4-1BB ligand receptor A930040I11Rik AA408498 AI325004 Cd137 CD137 antigen CDw137 EGK_00183 EGM_00177 homolog of mouse 4-1BB hypothetical protein ILA induced by lymphocyte activation (ILA) interleukin-activated receptor homolog of mouse Ly63 Ly63 receptor protein 4-1BB s4 1 BB s4 1BB s41BB sCD137 secreted CD137 antigen soluble 4 1 BB soluble 4 1BB soluble 41BB soluble CD137 T cell antigen ILA T-cell antigen 4-1BB T-cell antigen 4-1BB homolog T-cell antigen ILA TNF receptor superfamily member 9 Tnfrsf9 Tumor necrosis factor receptor superfamily member 9 tumor necrosis factor receptor superfamily member 9; 4-1BB ligand receptor; CD137; CD137 antigen; CDw137; homolog of mouse 4-1BB; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; receptor protein 4-1BB; T cell antigen ILA; T-cell antigen 4-1BB homolog; T-cell antigen ILA; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9
Gene Aliases: 4-1BB; CD137; CDw137; ILA; TNFRSF9
UniProt ID: (Human) Q07011
Entrez Gene ID: (Human) 3604
Molecular Function:
cytokine receptor
receptor
tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.