Invitrogen
Manufacturer Code:17137182
Catalog # 50-112-9043
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Syrian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | 17B5 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Syrian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: 4-1BB ligand receptor; 4-1BB ligand receptor CD137 antigen homolog of mouse 4-1BB induced by lymphocyte activation (ILA) interleukin-activated receptor homolog of mouse Ly63 receptor protein 4-1BB T cell antigen ILA T-cell antigen 4-1BB homolog T-cell antigen ILA tumor necrosis factor receptor superfamily member 9 tumor necrosis factor receptor superfamily member 9; CD137; CD137 antigen; secreted CD137 antigen; T-cell antigen 4-1BB; Tumor necrosis factor receptor superfamily member 9
Gene Aliases: 4-1BB; A930040I11Rik; AA408498; AI325004; Cd137; CDw137; ILA; Ly63; Tnfrsf9
UniProt ID: (Mouse) P20334
Entrez Gene ID: (Mouse) 21942
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.