Invitrogen
Manufacturer Code:13132182
Catalog # 50-245-545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | TUGm2 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD132; Cytokine receptor common subunit gamma; gammaC; IL-2 receptor subunit gamma; IL2RG CIDX IL-2RG IMD4 P64 SCIDX SCIDX1 CD132 antigen IL-2 receptor subunit gamma IL-2R subunit gamma cytokine receptor common subunit gamma gamma (c) gammaC interleukin-2 receptor subunit gamma IL2R gamma; Interleukin-2 receptor subunit gamma; p64
Gene Aliases: [g]c; CD132; gamma(c); gc; Il2rg; p64
UniProt ID: (Mouse) P34902
Entrez Gene ID: (Mouse) 16186
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.