Invitrogen
Manufacturer Code:MA119704
Catalog # 01-676-729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | WM15 |
Immunogen | Human AML cells. |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alanyl (membrane) aminopeptidase; Alanyl aminopeptidase; Aminopeptidase M; Aminopeptidase N; AP-M; AP-M AP-N Apn Cd13 P150 alanyl aminopeptidase aminopeptidase M aminopeptidase N aminopeptidase N/CD13 membrane protein p161 microsomal aminopeptidase; AP-N; CD13; gp150; hAPN; membrane alanyl aminopeptidase; Microsomal aminopeptidase; Myeloid plasma membrane glycoprotein CD13
Gene Aliases: ANPEP; APN; CD13; GP150; LAP1; P150; PEPN
UniProt ID: (Human) P15144
Entrez Gene ID: (Human) 290
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.