Proteintech
Manufacturer Code:145531AP150UL
Catalog # 50-172-9538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CD13 Fusion Protein Ag5976 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alanyl (membrane) aminopeptidase; Alanyl aminopeptidase; Alanyl aminopeptidase Aminopeptidase M Aminopeptidase N ANPEP AP M AP N APN CD13 gp150 hAPN LAP1 Microsomal aminopeptidase p150 PEPN; Aminopeptidase M; Aminopeptidase N; aminopeptidase N/CD13; AP-M; AP-N; CD13; cluster of differentiation antigen 13 (CD13); gp150; hAPN; kidney aminopeptidase M; Kidney Zn peptidase; KZP; leucine arylaminopeptidase 1; membrane alanine aminopeptidase; membrane alanyl aminopeptidase; Membrane protein p161; Microsomal aminopeptidase; Myeloid plasma membrane glycoprotein CD13
Gene Aliases: ANPEP; AP-M; AP-N; Apm; APN; CD13; GP150; KZP; Lap-1; LAP1; P150; PEPN; rAPN
UniProt ID: (Human) P15144, (Mouse) P97449, (Rat) P15684
Entrez Gene ID: (Human) 290, (Mouse) 16790, (Rat) 81641
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.