Invitrogen
Manufacturer Code:50127182
Catalog # 50-112-4301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | A7R34 |
Conjugate | eFluor 660 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD127; CD127 IL-7Ralpha MGC107557; IL-7 receptor alpha chain; IL-7 receptor subunit alpha; IL-7R subunit alpha; IL-7R-alpha; IL-7RA; interleukin 7 receptor alpha chain; Interleukin-7 receptor subunit alpha
Gene Aliases: CD127; IL-7Ralpha; Il7r
UniProt ID: (Mouse) P16872
Entrez Gene ID: (Mouse) 16197
Molecular Function:
cytokine receptor
receptor
type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.