Invitrogen
Manufacturer Code:MA529369
Catalog # PIMA529369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombit Human IL7R/IL-7R/CD127 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD127; CD127 antigen; CDw127; IL-7 receptor subunit alpha; IL-7R subunit alpha; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin-7 Receptor alpha CD127; Interleukin-7 receptor subunit alpha
Gene Aliases: CD127; CDW127; IL-7R-alpha; IL7R; IL7RA; ILRA
UniProt ID: (Human) Q6NWM0
Entrez Gene ID: (Human) 3575
Molecular Function:
cytokine receptor
receptor
type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.