Invitrogen
Manufacturer Code:PA5115054
Catalog # PIPA5115054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CSF2RA corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD116; CD116 antigen; CD116 CD116 antigen CDw116 colony stimulating factor 2 receptor alpha chain colony stimulating factor 2 receptor alpha subunit colony stimulating factor 2 receptor alpha low-affinity (granulocyte-macrophage) CSF 2R CSF2R CSF2RA CSF2RAX CSF2RAY CSF2RX CSF2RY Csfgmra GM CSF R alpha GM-CSF receptor alpha subunit GMCSFR GM-CSFR GM-CSF-Ra GM-CSFRalpha GM-CSF-R-alpha GMCSFR-alpha GMR GMR-alpha granulocyte-macrophage colony stimulating receptor alpha granulocyte-macrophage colony-stimulating factor receptor alpha chain granulocyte-macrophage colony-stimulating factor receptor subunit alpha MGC3848 MGC4838 SMDP4; CDw116; colony stimulating factor 2 receptor alpha chain; colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage); GM-CSF receptor alpha subunit; GM-CSF-R-alpha; GM-CSFRalpha; GMCSFR-alpha; GMR-alpha; granulocyte-macrophage colony stimulating receptor alpha; granulocyte-macrophage colony-stimulating factor receptor alpha chain; Granulocyte-macrophage colony-stimulating factor receptor subunit alpha
Gene Aliases: CD116; CDw116; CSF2R; CSF2RA; CSF2RAX; CSF2RAY; CSF2RX; CSF2RY; Csfgmra; GM-CSF-R-alpha; GM-CSF-Ra; GM-CSFR; GMCSFR; GMR; SMDP4
UniProt ID: (Human) P15509, (Mouse) Q00941
Entrez Gene ID: (Human) 1438, (Mouse) 12982, (Rat) 652957
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.