Novus Biologicals
Manufacturer Code:NBP275969
Catalog # NBP275969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | W7C5 |
Immunogen | WERI-RB-1 retinoblastoma cell line |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: 150 kDa TGF-beta-1-binding protein; 150 kDa TGF-beta-1-binding protein activated T-cell marker CD109 C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7 CD109 antigen CD109 antigen (Gov platelet alloantigens) CD109 molecule CPAMD7r150 DKFZp762L1111 FLJ38569 FLJ41966 Gov platelet alloantigens p180 Platelet-specific Gov antigen RP11-525G3.1; activated T-cell marker CD109; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; CD109; CD109 antigen; Gov platelet alloantigens; p180; Platelet-specific Gov antigen; r150
Gene Aliases: CD109; CPAMD7; p180; r150
UniProt ID: (Human) Q6YHK3
Entrez Gene ID: (Human) 135228
Molecular Function: complement component cytokine defense/immunity protein enzyme modulator protease inhibitor serine protease inhibitor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.