Invitrogen
Manufacturer Code:14109995
Catalog # 14-109-995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HU17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 150 kDa TGF-beta-1-binding protein; 150 kDa TGF-beta-1-binding protein 9930012E15Rik activated T-cell marker CD109 AI480638 C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7 Cd109 CD109 antigen CD109 molecule CPAMD7 GARP Gov platelet alloantigens GPI-anchored alpha 2 macroglobulin-related protein GPI-anchored alpha-2 macroglobulin-related protein p180 Platelet-specific Gov antigen r150 RP11-525G3.1; activated T-cell marker CD109; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; CD109; CD109 antigen; Gov platelet alloantigens; p180; Platelet-specific Gov antigen; r150
Gene Aliases: CD109; CPAMD7; p180; r150
UniProt ID: (Human) Q6YHK3
Entrez Gene ID: (Human) 135228
Molecular Function: complement component cytokine defense/immunity protein enzyme modulator protease inhibitor serine protease inhibitor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.