Invitrogen
Manufacturer Code:MA524208
Catalog # PIMA524208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 496920 |
Immunogen | Mouse myeloma cell line NS0-derived recombit human CD109 Val22-Ser1268 (Tyr703Ser and Thr1241Met) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 150 kDa TGF-beta-1-binding protein; activated T-cell marker CD109; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; CD109; CD109 antigen; CPAMD7 RP11-525G3.1 150 kDa TGF-beta-1-binding protein; Gov platelet alloantigens; p180; Platelet-specific Gov antigen; r150
Gene Aliases: CD109; CPAMD7; p180; r150
UniProt ID: (Human) Q6YHK3
Entrez Gene ID: (Human) 135228
Molecular Function: complement component cytokine defense/immunity protein enzyme modulator protease inhibitor serine protease inhibitor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.