Invitrogen
Manufacturer Code:11107942
Catalog # 50-112-9389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | eBioH4A3 |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD107 antigen-like family member A; CD107a; CD107a LAMPA LGP120 CD107 antigen-like family member A LAMP-1 lysosome-associated membrane glycoprotein 1 lysosome-associated membrane protein 1; LAMP-1; lysosomal-associated membrane protein 1; Lysosome-associated membrane glycoprotein 1; lysosome-associated membrane protein 1
Gene Aliases: CD107a; LAMP1; LAMPA; LGP120
UniProt ID: (Human) P11279
Entrez Gene ID: (Human) 3916
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.