Invitrogen
Manufacturer Code:14107937
Catalog # 14-107-937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | eBioH4A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 120 kDa lysosomal membrane glycoprotein AI196048 CD107 antigen-like family member A CD107a I79_011073 Lamp I Lamp1 LAMP-1 LAMPA LGP120 LGP-120 Lgpa LGP-A lysosomal associated membrane protein 1 Lysosomal associated membrane protein 1 (120 kDa) lysosomal membrane glycoprotein 1 lysosomal membrane glycoprotein A lysosomal-associated membrane protein 1 lysosome-associated membrane glycoprotein 1 LYSOSOME-ASSOCIATED MEMBRANE GLYCOPROTEIN 1 PRECURSOR (LAMP-1) (LGP-A) (LGP-120) (CD107A) (P2B) lysosome-associated membrane protein 1 P2B; CD107 antigen-like family member A; CD107a; LAMP-1; lysosomal-associated membrane protein 1; Lysosome-associated membrane glycoprotein 1; lysosome-associated membrane protein 1
Gene Aliases: CD107a; LAMP1; LAMPA; LGP120
UniProt ID: (Human) P11279
Entrez Gene ID: (Human) 3916
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.