Invitrogen
Manufacturer Code:46107182
Catalog # 50-160-00
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | eBio1D4B (1D4B) |
Conjugate | PerCP-eFluor 710 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: 120 kDa lysosomal membrane glycoprotein; CD107 antigen-like family member A; CD107a; CD107a LAMPA LGP120 CD107 antigen-like family member A LAMP-1 lysosome-associated membrane glycoprotein 1 lysosome-associated membrane protein 1; Lamp I; LAMP-1; LGP-120; LGP-A; lysosomal membrane glycoprotein 1; Lysosomal membrane glycoprotein A; Lysosome-associated membrane glycoprotein 1; LYSOSOME-ASSOCIATED MEMBRANE GLYCOPROTEIN 1 PRECURSOR (LAMP-1) (LGP-A) (LGP-120) (CD107A) (P2B); lysosome-associated membrane protein 1; P2B
Gene Aliases: AI196048; CD107a; Lamp-1; Lamp1
UniProt ID: (Mouse) P11438
Entrez Gene ID: (Mouse) 16783
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.