Invitrogen
Manufacturer Code:PA579203
Catalog # PIPA579203
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse CD105 recombit protein (Position: E27-L226). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD105; Cell surface MJ7/18 antigen; Endoglin; Endoglin Cell surface MJ7/18 antigen CD105 Eng Edg; transmembrane glycoprotein
Gene Aliases: AI528660; AI662476; CD105; Edg; Endo; Eng; S-endoglin
UniProt ID: (Mouse) Q63961
Entrez Gene ID: (Mouse) 13805, (Rat) 497010
Molecular Function:
TGF-beta receptor
cytokine receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.