Invitrogen
Manufacturer Code:PA546971
Catalog # PIPA546971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Endoglin/CD105 Glu27-Gly581 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD105; CD105 END HHT1 ORW ORW1 CD105 antigen ENG; Cell surface MJ7/18 antigen; Endoglin; transmembrane glycoprotein
Gene Aliases: AI528660; AI662476; CD105; Edg; Endo; Eng; S-endoglin
UniProt ID: (Mouse) Q63961
Entrez Gene ID: (Mouse) 13805
Molecular Function: TGF-beta receptor cytokine receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.