Novus Biologicals
Manufacturer Code:NBP243680AF532
Catalog # NP243680A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 639 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human TCP1 epsilon. The exact sequence is proprietary. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CCT-epsilon; chaperonin containing TCP1 subunit 5 (epsilon) epsilon subunit; chaperonin containing TCP1, subunit 5 (epsilon); epididymis secretory protein Li 69; T-complex protein 1 subunit epsilon; TCP-1-epsilon
Gene Aliases: CCT-epsilon; CCT5; CCTE; HEL-S-69; KIAA0098; PNAS-102; TCP-1-epsilon
UniProt ID: (Human) P48643
Entrez Gene ID: (Human) 22948
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.