Proteintech
Manufacturer Code:116031AP150UL
Catalog # 50-172-7301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CCT5 Fusion Protein Ag2146 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCT epsilon CCT5 CCTE KIAA0098 TCP 1 epsilon; CCT-epsilon; chaperonin containing TCP1, subunit 5 (epsilon); chaperonin subunit 5 (epsilon); epididymis secretory protein Li 69; T-complex protein 1 subunit epsilon; TCP-1 epsilon; TCP-1-epsilon; unm tf212
Gene Aliases: CCT-epsilon; CCT5; CCTE; HEL-S-69; KIAA0098; mKIAA0098; PNAS-102; TCP-1-epsilon; TCPE; unm_tf212; wu:fb54h08
UniProt ID: (Human) P48643, (Mouse) P80316, (Rat) Q68FQ0
Entrez Gene ID: (Human) 22948, (Mouse) 12465, (Rat) 294864, (Zebrafish) 322258
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.