Invitrogen
Manufacturer Code:PA5117055
Catalog # PIPA5117055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CCT3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AL024092 Cct3 Cctg CCT-gamma chaperonin containing TCP1 subunit 3 chaperonin containing Tcp1 subunit 3 (gamma) chaperonin subunit 3 (gamma) hTRiC5 Matricin mTRiC-P5 OTTHUMP00000025735 PIG48 T-complex protein 1 subunit gamma T-complex protein 1 gamma subunit TCP1 (t-complex-1) ring complex polypeptide 5 TCP-1-gamma Tcp1-rs3 TRIC5 TriC-P5; CCT-gamma; chaperonin containing TCP1, subunit 3 (gamma); chaperonin subunit 3 (gamma); hTRiC5; Matricin; mTRiC-P5; T-complex protein 1 subunit gamma; T-complex protein 1, gamma subunit; TCP-1-gamma; TCP1 (t-complex-1) ring complex, polypeptide 5
Gene Aliases: AL024092; CCT-gamma; CCT3; CCTG; PIG48; TCP-1-gamma; Tcp1-rs3; TriC-P5; TRIC5
UniProt ID: (Human) P49368, (Mouse) P80318, (Rat) Q6P502
Entrez Gene ID: (Human) 7203, (Mouse) 12462, (Rat) 295230
Molecular Function:
chaperone
chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.