Abnova
Manufacturer Code:MAB12595
Catalog # 89-933-737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | PK/8/4/4i/2f |
Immunogen | Mouse Cct2 peptide-PPD conjugated. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against mouse Cct2.
For Research Use Only
Protein Aliases: CCT-beta; Cctb; chaperonin subunit 2 (beta); T-complex protein 1 subunit beta; TCP-1-beta
Gene Aliases: Cct2; Cctb
UniProt ID: (Mouse) P80314
Entrez Gene ID: (Mouse) 12461
Molecular Function:
chaperone
chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.